p-Tolyl trifluoromethanesulphonate ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Methylphenyl Trifluoromethanesulphonate, Trifluoromethanesulphonic acid p-Tolyl Ester, p-Tolyl Triflate

T2606-5G
TCIAT2606-5GEA 796 DKK
TCIAT2606-5G
p-Tolyl trifluoromethanesulphonate ≥95.0% (by GC)
p-Tolyl trifluoromethanesulphonate
Formel: C₈H₇F₃O₃S
Kogepunkt: 115°C
Flammepunkt: 89°C
MDL Number: MFCD00192507
CAS-nummer: 29540-83-8

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR