Ditetradecyl-3,3'-thiodipropionate ≥94.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3,3'-Thiodipropionic Acid Ditetradecyl Ester, 3,3'-Thiodipropionic Acid Dimyristyl Ester, Dimyristyl 3,3'-Thiodipropionate, 3,3-Thiodipropionicaciddimyristylester

T0911-25G T0911-500G
TCIAT0911-25GEA 132 DKK
TCIAT0911-25G TCIAT0911-500G
Ditetradecyl-3,3'-thiodipropionate ≥94.0% (by GC)
Ditetradecyl-3,3'-thiodipropionate
Formel: C₃₄H₆₆O₄S
MDL Number: MFCD00048462
CAS-nummer: 16545-54-3
EINECS: 240-613-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 94.0 %
Melting point 49.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR