Chinolin ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Q0085-25G Q0085-500G
TCIAQ0085-25GEA 197 DKK
TCIAQ0085-25G TCIAQ0085-500G
Chinolin ≥97.0% (by GC)
Quinoline
Formel: C₉H₇N
Kogepunkt: 238°C
Smeltepunkt: –15°C
Massefylde: 1,095 g/cm³ (20 °C)
Flammepunkt: 106°C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006736
CAS-nummer: 91-22-5
EINECS: 202-051-6
UN nummer: 2656
ADR: 6.1,III
Merck Index: 14,08068
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=158670e9b21d838d9ea9c4f43510f7ba095f5f2d&vid=cbc3fd17586f088699d1dc82ee081d62c07bb462&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR