[6,6]-Phenyl-C61-butyric acid methyl ester ≥99.5% (by HPLC) for organic electronics

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: [60]PCBM, PCBM, [6,6]-Phenyl-C61-butyric Acid Methyl Ester, 3′H-Cyclopropa[1,9] [5,6]fullerene-C60-Ih-3′-butanoic acid 3′-phenyl methyl ester, Methyl [6,6]-Phenyl-C61-butyrate

P2682-100MG
TCIAP2682-100MGEA 1990 DKK
TCIAP2682-100MG
[6,6]-Phenyl-C61-butyric acid methyl ester ≥99.5% (by HPLC) for organic electronics
[6,6]-Phenyl-C61-butyric acid methyl ester
Formel: C₇₂H₁₄O₂
MDL Number: MFCD07784544
CAS-nummer: 160848-22-6

Bestil nu

Specifikationer Testresultater

Appearance Dark green to Dark red to Black powder to crystal
Purity (HPLC) min. 99.5 area%
Electron Mobility (muFET) min. 0.020 cm2/Vs (HMDS Si/SiO2 substrate)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR