Phytantriol (mixture of isomers) ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3,7,11,15-Tetramethylhexadecane-1,2,3-triol, 1,2,3-Trihydroxy-3,7,11,15-tetramethylhexadecane

P1674-5G
TCIAP1674-5GEA 2150 DKK
TCIAP1674-5G
Phytantriol (mixture of isomers) ≥95.0% (by GC)
Phytantriol (mixture of isomers)
Formel: C₂₀H₄₂O₃
Molekylvægt: 330.55 g/mol
Flammepunkt: 199°C
CAS-nummer: 74563-64-7
EINECS: 277-923-2
Merck Index: 14,07386

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 % (total of isomers)
Purity (Hydroxyl Value) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR