Pentadecanenitrile ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Myristyl cyanide, Tetradecyl cyanide, Pentadecanonitrile

P0871-25G P0871-5G
TCIAP0871-25GEA 2060 DKK
TCIAP0871-25G TCIAP0871-5G
Pentadecanenitrile ≥98.0% (by GC)
Pentadecanenitrile
Formel: C₁₅H₂₉N
MDL Number: MFCD00019849
CAS-nummer: 18300-91-9
EINECS: 242-184-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=911363b0e51a0a5943174cd0b1f917e51571504f&vid=911363b0e51a0a5943174cd0b1f917e51571504f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR