2-Hydroxyethyl phenyl sulphide ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(Phenylthio)ethanol

P0733-25ML P0733-500ML
TCIAP0733-25MLEA 255 DKK
TCIAP0733-25ML TCIAP0733-500ML
2-Hydroxyethyl phenyl sulphide ≥97.0% (by GC)
2-Hydroxyethyl phenyl sulphide
Formel: C₈H₁₀OS
MDL Number: MFCD00002905
CAS-nummer: 699-12-7
EINECS: 211-828-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b171267d30f3b3f41d756d76356a933cba7a38a&vid=4b171267d30f3b3f41d756d76356a933cba7a38a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR