(±)-1-Phenyl-1-propanol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Phenyl-1-propanol, (±)-α-Ethylbenzyl alcohol

P0212-25G P0212-500G
TCIAP0212-25GEA 189 DKK
TCIAP0212-25G TCIAP0212-500G
(±)-1-Phenyl-1-propanol ≥98.0% (by GC)
(±)-1-Phenyl-1-propanol
Formel: C₆H₅CH(OH)CH₂CH₃
Kogepunkt: 212°C
Massefylde: 0,99 g/cm³ (20 °C)
Flammepunkt: 90°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004564
CAS-nummer: 93-54-9
EINECS: 202-256-0
Merck Index: 14,03768
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1176d449df743790a4706d5ab04c6637d1d34a55&vid=1176d449df743790a4706d5ab04c6637d1d34a55&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR