4-Nitrobenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

N0989-5G N0989-25G
TCIAN0989-5GEA 485 DKK
TCIAN0989-5G TCIAN0989-25G
4-Nitrobenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Nitrobenzamidine hydrochloride
Formel: C₇H₇N₃O₂
Opbevaringstemperatur: Fryser
MDL Number: MFCD00278833
CAS-nummer: 15723-90-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9ee37a28eeb672987159c1929f0e2e69102d756&vid=b9ee37a28eeb672987159c1929f0e2e69102d756&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Dark green powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR