Nonanal dimethyl acetal ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Pelargonaldehyde Dimethyl Acetal, 1,1-Dimethoxynonane, Nonyl Aldehyde Dimethyl Acetal

N0915-5G N0915-25G
TCIAN0915-5GEA 230 DKK
TCIAN0915-5G TCIAN0915-25G
Nonanal dimethyl acetal ≥97.0% (by GC)
Nonanal dimethyl acetal
Formel: C₁₁H₂₄O₂
Kogepunkt: 88°C
MDL Number: MFCD00036646
CAS-nummer: 18824-63-0
EINECS: 242-603-3

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR