Lithium perfluorobutane-1-sulphonate ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Lithium Nonafluoro-1-butanesulphonate, Lithium nonafluorobutanesulphonate

N0710-25G
TCIAN0710-25GEA 1070 DKK
TCIAN0710-25G
Lithium perfluorobutane-1-sulphonate ≥95.0% (by titrimetric analysis)
Lithium perfluorobutane-1-sulphonate
Formel: C₄F₉LiO₃S
Molekylvægt: 306.0326 g/mol
MDL Number: MFCD03094160
CAS-nummer: 131651-65-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=560a237cf3ab44495ac3fe91351ef5cb9695b608&vid=560a237cf3ab44495ac3fe91351ef5cb9695b608&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 95.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR