2-Nitro-p-toluonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Methyl-2-nitrobenzonitrile

N0494-1G N0494-5G
TCIAN0494-1GEA 657 DKK
TCIAN0494-1G TCIAN0494-5G
2-Nitro-p-toluonitril ≥98.0% (by GC)
2-Nitro-p-toluonitril
Formel: C₈H₆N₂O₂
MDL Number: MFCD00056107
CAS-nummer: 26830-95-5
EINECS: 248-019-5
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=457c33b35706b60abda7352d2bb27dd88c1f310a&vid=457c33b35706b60abda7352d2bb27dd88c1f310a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 100.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR