Ethyl 4-nitrobenzoat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Ethyl 4-nitrobenzoate, 4-Nitrobenzoic acid ethyl ester

N0485-25G
TCIAN0485-25GEA 263 DKK
TCIAN0485-25G
Ethyl 4-nitrobenzoat ≥98.0% (by GC)
Ethyl 4-nitrobenzoat
Formel: C₉H₉NO₄
Molekylvægt: 195.17 g/mol
Kogepunkt: 182°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007351
CAS-nummer: 99-77-4
EINECS: 202-786-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=188315e284d097274265d00113c1f8bdea7406cd&vid=188315e284d097274265d00113c1f8bdea7406cd&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Red powder to crystalline
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR