1,4-Naphthochinon ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1,4-Naphthalenedione, 1,4-Dihydro-1,4-diketonaphthalene

N0040-500G N0040-25G N0040-100G
TCIAN0040-500GEA 1100 DKK
TCIAN0040-500G TCIAN0040-25G TCIAN0040-100G
1,4-Naphthochinon ≥98.0% (by GC)
1,4-Naphthochinon
Formel: C₁₀H₆O₂
Kogepunkt: 298 °C (1013 hPa)
Smeltepunkt: 124 °C
Massefylde: 1,422 g/cm³ (20 °C)
Flammepunkt: 141 °C
MDL Number: MFCD00001676
CAS-nummer: 130-15-4
EINECS: 204-977-6
UN nummer: 2811
ADR: 6.1,I
Merck Index: 12,06482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3368db37bc7a7041b81f7b28843d42d684ce0af&vid=b3368db37bc7a7041b81f7b28843d42d684ce0af&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Cerium redox Method) min. 95.0 %
Melting point 123.0 to 126.0 deg-C
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR