2-Methyl-1H-indole-3-carbaldehyde ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Methylindole-3-carboxaldehyde, 3-Formyl-2-methylindole

M1485-25G M1485-5G
TCIAM1485-25GEA 1900 DKK
TCIAM1485-25G TCIAM1485-5G
2-Methyl-1H-indole-3-carbaldehyde ≥98.0% (by GC)
2-Methyl-1H-indole-3-carbaldehyde
Formel: C₁₀H₉NO
MDL Number: MFCD00012077
CAS-nummer: 5416-80-8
EINECS: 226-512-6

Bestil nu

Specifikationer Testresultater

Purity (GC) min. 98.0 %
Melting point 201.0 to 205.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR