N-Phenyl-m-toluidin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Methyldiphenylamine

M1258-25G
TCIAM1258-25GEA 312 DKK
TCIAM1258-25G
N-Phenyl-m-toluidin ≥98.0% (by GC)
N-Phenyl-m-toluidin
Formel: C₁₃H₁₃N
Molekylvægt: 183.25 g/mol
MDL Number: MFCD00008530
CAS-nummer: 1205-64-7
EINECS: 214-885-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d146966a07d3d99b0020119dd73fdb9e63670428&vid=d146966a07d3d99b0020119dd73fdb9e63670428&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR