Nitro-tert-butan ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Methyl-2-nitropropane

M1241-5G
TCIAM1241-5GEA 296 DKK
TCIAM1241-5G
Nitro-tert-butan ≥98.0% (by GC)
Nitro-tert-butan
Formel: C₄H₉NO₂
Kogepunkt: 126 °C (1013 hPa)
Massefylde: 1,059 g/cm³ (20 °C)
Flammepunkt: 19°C
MDL Number: MFCD00007393
CAS-nummer: 594-70-7
EINECS: 209-851-4
UN nummer: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09c517a139791ab8145128591a9061d91bab7260&vid=76876306fe6c79b0ea94040964c9f8d941f7ae94&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR