Ethyl-O-Mesitylsulphonylacetohydroxamate ≥98.0% (by HPLC, total nitrogen) precursor of the powerful aminating reagent

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: O-Mesitylsulphonylacetohydroxamic acid Ethyl Ester, Ethyl N-(mesitylsulphonyl)oxyacetimidate

M1182-1G M1182-25G M1182-5G
TCIAM1182-1GEA 132 DKK
TCIAM1182-1G TCIAM1182-25G TCIAM1182-5G
Ethyl-O-Mesitylsulphonylacetohydroxamate ≥98.0% (by HPLC, total nitrogen) precursor of the powerful aminating reagent
Ethyl-O-Mesitylsulphonylacetohydroxamate
Formel: C₁₃H₁₉NO₄S
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00009244
CAS-nummer: 38202-27-6
EINECS: 253-825-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 54.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR