Lithiumtert-butanolat ∼10% ∼1 mol/L in THF

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L0253-100ML
TCIAL0253-100MLEA 862 DKK
TCIAL0253-100ML
Lithiumtert-butanolat ∼10% ∼1 mol/L in THF
Lithiumtert-butanolat
Formel: C₄H₉LiO
Massefylde: 0,89
MDL Number: MFCD00050479
CAS-nummer: 1907-33-1
EINECS: 217-611-5
UN nummer: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da24e69d234890b06b5fb12dd031ed7ea1bfaa47&vid=669c2b76e33cea6e27d87e14f0b29a496724e416&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown clear liquid
Concentration (Neutralization titration) 9.6 to 10.2 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR