Blytetraacetat ≥96.0% (by titrimetric analysis) (contains Acetic Acid)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Lead(IV) tetraacetate, Triacetyloxyplumbyl acetate, Acetic acid lead (IV) salt, Lead (IV) acetate, LTA

L0021-25G L0021-500G
TCIAL0021-25GEA 435 DKK
TCIAL0021-25G TCIAL0021-500G
Blytetraacetat ≥96.0% (by titrimetric analysis) (contains Acetic Acid)
Blytetraacetat
Formel: (H₃CCOO)₄Pb
Smeltepunkt: 175…180 °C
Massefylde: ~ 2,22 g/cm³ (20 °C)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00008693
CAS-nummer: 546-67-8
EINECS: 208-908-0
UN nummer: 3087
ADR: 5.1,III
Merck Index: 14,05423
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d2baa2454f3fed03ec227df6aad5378dd3cff5c&vid=99bc5581abd632c1308d35bbc90594d65816329b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR