4-iod-2-nitroanilin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

I1029-1G I1029-5G
TCIAI1029-1GEA 206 DKK
TCIAI1029-1G TCIAI1029-5G
4-iod-2-nitroanilin ≥98.0% (by GC)
4-iod-2-nitroanilin
Formel: C₆H₅IN₂O₂
MDL Number: MFCD06808526
CAS-nummer: 20691-72-9

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 124.0 to 128.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR