4-Iodtoluen ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Iodo-4-methylbenzene

I0218-25G I0218-500G I0218-100G
TCIAI0218-25GEA 279 DKK
TCIAI0218-25G TCIAI0218-500G TCIAI0218-100G
4-Iodtoluen ≥99.0% (by GC)
4-Iodtoluen
Formel: C₇H₇I
Kogepunkt: 211,5 °C (1013 hPa)
Smeltepunkt: 33…35 °C
Massefylde: 1,68 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001059
CAS-nummer: 624-31-7
EINECS: 210-841-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbaf4b78f4735e6c110e8c400a4167161f41e112&vid=9933bb546727ceb451bb694466c8b4d6f4159763&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 99.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR