Isobutyl sulphide ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Diisobutyl Sulphide

I0098-25ML
TCIAI0098-25MLEA 411 DKK
TCIAI0098-25ML
Isobutyl sulphide ≥98.0% (by GC)
Isobutyl sulphide
Formel: C₈H₁₈S
Kogepunkt: 170°C
Flammepunkt: 52°C
MDL Number: MFCD00039852
CAS-nummer: 592-65-4
UN nummer: 1993
ADR: 3,III
Merck Index: 14,05152

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR