Hydrogenchlorid ∼4 mol/L 1,4-dioxane

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H1062-25ML H1062-500ML H1062-100ML
TCIAH1062-25MLEA 173 DKK
TCIAH1062-25ML TCIAH1062-500ML TCIAH1062-100ML
Hydrogenchlorid ∼4 mol/L 1,4-dioxane
Hydrogen chloride
Formel: HCl
Molekylvægt: 36.46 g/mol
Flammepunkt: 17°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
UN nummer: 2924
ADR: 3 II
Merck Index: 14,04780
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=338900896177ed872f84260a2c7a1dc5ecd792bb&vid=e5980c0cffb1dc3880f1220a8899149e8f4ae26c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Concentration (Neutralization titration) 14.0 to 15.4 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR