HEPES ≥99.0% (by titrimetric analysis) good's buffer component for biological research

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

H0396-25G H0396-500G
TCIAH0396-25GEA 132 DKK
TCIAH0396-25G TCIAH0396-500G
HEPES ≥99.0% (by titrimetric analysis) good's buffer component for biological research
HEPES
Formel: C₈H₁₈N₂O₄S
Smeltepunkt: 210…215 °C
Massefylde: 1,448 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006158
CAS-nummer: 7365-45-9
EINECS: 230-907-9
Merck Index: 14,04654
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5712d9b5cbff92ce8695d879fb0c0bfe0f0c011&vid=f5712d9b5cbff92ce8695d879fb0c0bfe0f0c011&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %
Absorbance max. 0.25 (1mol/L, H2O, 250nm)
Absorbance2 max. 0.15 (1mol/L、H2O、260nm)
Absorbance3 max. 0.10 (1mol/L、H2O、320nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR