(R)-(+)-2,3-Epoxy-1-propanol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (R)-(+)-2-(Hydroxymethyl)oxirane, (R)-(+)-1-Hydroxy-2,3-epoxypropane, (R)-Oxiran-2-ylmethanol, (R)-(+)-Glycidol

G0363-5G
TCIAG0363-5GEA 649 DKK
TCIAG0363-5G
(R)-(+)-2,3-Epoxy-1-propanol ≥98.0% (by GC)
(R)-(+)-2,3-Epoxy-1-propanol
Formel: C₃H₆O₂
Kogepunkt: 57°C
Flammepunkt: 72°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00074873
CAS-nummer: 57044-25-4
EINECS: 404-660-4
UN nummer: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be2bb5e918068ad6e343b995cbc090c7b5669e85&vid=be2bb5e918068ad6e343b995cbc090c7b5669e85&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%
Specific rotation [a]20/D +13.0 to +17.0 deg (neat)

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR