Geranyl acetate ≥70% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (E)-3,7-Dimethylocta-2,6-dien-1-yl acetate

G0028-25ML G0028-500ML G0028-100ML
TCIAG0028-25MLEA 165 DKK
TCIAG0028-25ML TCIAG0028-500ML TCIAG0028-100ML
Geranyl acetate ≥70% (by GC)
Geranyl acetate
Formel: C₁₂H₂₀O₂
Kogepunkt: 120°C
Flammepunkt: 118°C
MDL Number: MFCD00015037
CAS-nummer: 105-87-3
EINECS: 203-341-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=467f1607f7a97e2912b68265384ae08539b51c95&vid=467f1607f7a97e2912b68265384ae08539b51c95&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 70.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR