1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Vinyl-7-oxabicyclo[4.1.0]heptane, 4-Vinyl-1,2-epoxycyclohexane

E0956-25ML E0956-250ML
TCIAE0956-25MLEA 148 DKK
TCIAE0956-25ML TCIAE0956-250ML
1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) ≥98.0% (by GC)
1,2-Epoxy-4-vinylcyclohexane (mixture of isomers)
Formel: C₈H₁₂O
Kogepunkt: 159°C
Smeltepunkt: -100°C
Flammepunkt: 58°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00022356
CAS-nummer: 106-86-5
EINECS: 203-436-1
UN nummer: 3271
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR