Etodolac ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Etodlic acid, 1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid, 2-(1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl)acetic acid

E0858-25G E0858-5G
TCIAE0858-25GEA 1140 DKK
TCIAE0858-25G TCIAE0858-5G
Etodolac ≥98.0% (by HPLC, titration analysis)
Etodolac
Formel: C₁₇H₂₁NO₃
Smeltepunkt: 148°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00133313
CAS-nummer: 41340-25-4
UN nummer: 2811
ADR: 6.1,III
Merck Index: 14,03874

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR