Ethyl (2,3,4,5-tetrafluorobenzoyl)acetate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (2,3,4,5-Tetrafluorobenzoyl)acetic Acid Ethyl Ester, Ethyl 3-Oxo-3-(2,3,4,5-tetrafluorophenyl)propionate, Ethyl 3-oxo-3-(2,3,4,5-tetrafluorophenyl)propanoate, 3-Oxo-3-(2,3,4,5-tetrafluorophenyl)propionic Acid Ethyl Ester

E0759-25G
TCIAE0759-25GEA 591 DKK
TCIAE0759-25G
Ethyl (2,3,4,5-tetrafluorobenzoyl)acetate ≥98.0% (by GC)
Ethyl (2,3,4,5-tetrafluorobenzoyl)acetate
Formel: C₁₁H₈F₄O₃
Kogepunkt: 100°C
MDL Number: MFCD00523020
CAS-nummer: 94695-50-8

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0% (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR