Ethylbenzen ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Phenylethane

E0064-25ML E0064-500ML
TCIAE0064-25MLEA 132 DKK
TCIAE0064-25ML TCIAE0064-500ML
Ethylbenzen ≥99.0% (by GC)
Ethylbenzen
Formel: C₈H₁₀
Kogepunkt: 136°C
Smeltepunkt: –95 °C
Massefylde: 0,867 g/cm³ (20 °C)
Flammepunkt: 25°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011647
CAS-nummer: 100-41-4
EINECS: 202-849-4
UN nummer: 1175
ADR: 3,II
Merck Index: 14,03765
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2836d611086b5b60227a4ef9c3b1e03131835549&vid=22fc297aa5000d7e3278522aa4b1b4bd689f1df0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR