1,2-Ethandithiol ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1.2-Ethanedithiol, 1,2-Dimercaptoethane

E0032-25G E0032-500G E0032-100G
TCIAE0032-25GEA 238 DKK
TCIAE0032-25G TCIAE0032-500G TCIAE0032-100G
1,2-Ethandithiol ≥99.0% (by GC)
1,2-Ethandithiol
Formel: C₂H₆S₂
Kogepunkt: 146°C
Smeltepunkt: –41 °C
Massefylde: 1,374 g/cm³ (20 °C)
Flammepunkt: 44°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004892
CAS-nummer: 540-63-6
EINECS: 208-752-3
UN nummer: 3336
ADR: 3,III
Merck Index: 14,03725
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6257afcd312f382856807a7b4be209a0eb0be286&vid=7bc97f63b7b71ed1798c689524f7cde13d0f53b5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR