2,7-Dibromophenanthrene-9,10-dione ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D4363-1G D4363-5G
TCIAD4363-1GEA 427 DKK
TCIAD4363-1G TCIAD4363-5G
2,7-Dibromophenanthrene-9,10-dione ≥97.0% (by GC)
2,7-Dibromophenanthrene-9,10-dione
Formel: C₁₄H₆Br₂O₂
Smeltepunkt: 331°C
MDL Number: MFCD03931078
CAS-nummer: 84405-44-7

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR