1,3-Dimethoxy-1,3-dimethylurea ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D3990-5G
TCIAD3990-5GEA 1800 DKK
TCIAD3990-5G
1,3-Dimethoxy-1,3-dimethylurea ≥97.0% (by GC)
1,3-Dimethoxy-1,3-dimethylurea
Formel: C₅H₁₂N₂O₃
Kogepunkt: 42°C
Opbevaringstemperatur: Køleskab
CAS-nummer: 123707-26-6
UN nummer: 1993
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR