Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-on ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Dimethylpropylenurinstof

D2014-25G D2014-500G D2014-100G
TCIAD2014-25GEA 148 DKK
TCIAD2014-25G TCIAD2014-500G TCIAD2014-100G
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-on ≥98.0% (by GC)
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-on
Formel: C₆H₁₂N₂O
Kogepunkt: 93°C
Smeltepunkt: –24 °C
Massefylde: 1,06 g/cm³ (20 °C)
Flammepunkt: 121°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006550
CAS-nummer: 7226-23-5
EINECS: 230-625-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19b2f8411a27de13749a9ada44969026d54b311d&vid=19b2f8411a27de13749a9ada44969026d54b311d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR