N,N'-Dimethyl-3-aminopyrrolidine ≥96.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N,1-Dimethylpyrrolidin-3-amine

D1965-10G
TCIAD1965-10GEA 2040 DKK
TCIAD1965-10G
N,N'-Dimethyl-3-aminopyrrolidine ≥96.0% (by GC, titration analysis)
N,N'-Dimethyl-3-aminopyrrolidine
Formel: C₆H₁₄N₂
Molekylvægt: 114.19 g/mol
Kogepunkt: 167°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00059061
CAS-nummer: 64021-83-6
UN nummer: 2733
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80c105f96f7e4d2b325e62936e4750c6ca2c19f9&vid=80c105f96f7e4d2b325e62936e4750c6ca2c19f9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR