1,3-Dimethoxybenzen ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Resorcinol dimethyl ether

D0628-25G D0628-500G D0628-100G
TCIAD0628-25GEA 132 DKK
TCIAD0628-25G TCIAD0628-500G TCIAD0628-100G
1,3-Dimethoxybenzen ≥99.0% (by GC)
1,3-Dimethoxybenzen
Formel: C₈H₁₀O₂
Molekylvægt: 138.17 g/mol
Kogepunkt: 217°C
Smeltepunkt: -52
Flammepunkt: 87°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008384
CAS-nummer: 151-10-0
EINECS: 205-783-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=627503d26cad73a77d9ec52261edddf45fbb779f&vid=c5a513763aa252a5017bf3b2a2abc51910a360e0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR