Digitonin

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Digitonin

D0540-100MG
TCIAD0540-100MGEA 657 DKK
TCIAD0540-100MG
Digitonin
Digitonin
Formel: C₅₆H₉₂O₂₉
MDL Number: MFCD00077729
CAS-nummer: 11024-24-1
EINECS: 234-255-6
UN nummer: 2811
ADR: 6.1,III
Merck Index: 14,03161
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=beb2170db52c1125f15b045cefb84d0973f6f86e&vid=beb2170db52c1125f15b045cefb84d0973f6f86e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystaline
Specific rotation [a]20/D -49.0 to -57.0 deg (C=1, EtOH) (after drying)
Drying loss max. 8.0 %
Sensitiveness to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR