1,1-Dibromethan ≥98.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ethylidene bromide

Stabilization: Stabilised with copper chip

D0184-5G
TCIAD0184-5GEA 1020 DKK
TCIAD0184-5G
1,1-Dibromethan ≥98.0% (by GC) stabilised
1,1-Dibromoethane
Formel: Br₂HCCH₃
Molekylvægt: 187.86 g/mol
Kogepunkt: 108°C
Smeltepunkt: -0.5
MDL Number: MFCD00041719
CAS-nummer: 557-91-5
EINECS: 209-184-9
UN nummer: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a74aa3cb7275edbe3a93c25b17f5adc240f2983&vid=2a74aa3cb7275edbe3a93c25b17f5adc240f2983&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR