2-Chlorophenyl cyclopentyl ketone ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Chlorobenzoylcyclopentane

C2338-5G
TCIAC2338-5GEA 1460 DKK
TCIAC2338-5G
2-Chlorophenyl cyclopentyl ketone ≥98.0% (by GC)
2-Chlorophenyl cyclopentyl ketone
Formel: C₁₂H₁₃ClO
Kogepunkt: 96°C
MDL Number: MFCD00038367
CAS-nummer: 6740-85-8
EINECS: 229-802-0

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR