4-Chlor-2-methoxybenzoesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Chloro-o-anisic acid

C2076-25G C2076-5G
TCIAC2076-25GEA 911 DKK
TCIAC2076-25G TCIAC2076-5G
4-Chlor-2-methoxybenzoesyre ≥98.0% (by GC, titration analysis)
4-Chlor-2-methoxybenzoesyre
Formel: C₈H₇ClO₃
Kogepunkt: 308 °C (1013 hPa)
Smeltepunkt: 148 °C
MDL Number: MFCD00002532
CAS-nummer: 57479-70-6
EINECS: 260-761-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1925b6e0b40540d22570d253c7eadd3eeb28c8f1&vid=1925b6e0b40540d22570d253c7eadd3eeb28c8f1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency
Melting point 146.0 and le 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR