Vinylbenzyl cyanide (m and p mixture) ≥95.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Cyanomethylstyrene, Vinylphenylacetonitrile

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

C1281-5G
TCIAC1281-5GEA 1240 DKK
TCIAC1281-5G
Vinylbenzyl cyanide (m and p mixture) ≥95.0% (by GC) stabilised
Vinylbenzyl cyanide (m and p mixture)
Formel: C₁₀H₉N
Molekylvægt: 143.19 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00060668
UN nummer: 3276
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab3bc9dbf41ea82a7cc0495dced5e35f96ed5f2c&vid=14ab8ce91df695fa1af5bf4cf202066fb798182c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR