5-Chloro-2-methylaniline Hydrochloride ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Amino-4-chlorotoluene Hydrochloride, 5-Chloro-2-methylaniline hydrochloride, 5-Chloro-o-toluidine Hydrochloride

C0746-25G
TCIAC0746-25GEA 944 DKK
TCIAC0746-25G
5-Chloro-2-methylaniline Hydrochloride ≥98.0% (by HPLC, total nitrogen)
5-Chloro-2-methylaniline Hydrochloride
Formel: C₇H₈ClN
MDL Number: MFCD00035487
CAS-nummer: 6259-42-3
EINECS: 228-403-9

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR