Cyclooctanol 98,0 GC_ASSAY_METHOD

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C0623-25G
TCIAC0623-25GEA 690 DKK
TCIAC0623-25G
Cyclooctanol 98,0 GC_ASSAY_METHOD
Cyclooctanol
Formel: C₈H₁₆O
Molekylvægt: 128.21 g/mol
Kogepunkt: 108°C
Smeltepunkt: 15
Flammepunkt: 88°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001744
CAS-nummer: 696-71-9
EINECS: 211-800-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb7058612ca2d9515e4b0798eaa840a00f72092a&vid=eb7058612ca2d9515e4b0798eaa840a00f72092a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR