2-Chloroethyl methyl sulphide ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (2-Chloroethyl)(methyl)sulphane

C0588-10G
TCIAC0588-10GEA 772 DKK
TCIAC0588-10G
2-Chloroethyl methyl sulphide ≥98.0% (by GC)
2-Chloroethyl methyl sulphide
Formel: C₃H₇ClS
Kogepunkt: 140°C
Flammepunkt: 42°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00000978
CAS-nummer: 542-81-4
EINECS: 208-828-6
UN nummer: 2924
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2ac6dab0c9ecc65fa7c889e37929c9a7a5715a4&vid=e2ac6dab0c9ecc65fa7c889e37929c9a7a5715a4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR