2-Hydroxy-3-methylbenzoesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2,3-Cresotic acid, 2-Hydroxy-3-methylbenzoic acid, 3-Methylsalicylic acid

C0408-25G C0408-500G C0408-100G
TCIAC0408-25GEA 197 DKK
TCIAC0408-25G TCIAC0408-500G TCIAC0408-100G
2-Hydroxy-3-methylbenzoesyre ≥98.0% (by GC, titration analysis)
2-Hydroxy-3-methylbenzoesyre
Formel: C₈H₈O₃
Kogepunkt: 293 °C (1013 hPa)
Smeltepunkt: 163…165 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002448
CAS-nummer: 83-40-9
EINECS: 201-473-8
Merck Index: 14,02581
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c0b7497d9916d34b068de3c755c8715dd60d392&vid=4b765989dc5709263034098e24c32e229caf17d3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 165.0 to 169.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR