L-Canavanine sulphate ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Canavanine sulphate, L-α-Amino-γ-(guanidinooxy)-n-butyric acid sulphate

C0018-100MG
TCIAC0018-100MGEA 698 DKK
TCIAC0018-100MG
L-Canavanine sulphate ≥97.0% (by titrimetric analysis)
L-Canavanine sulphate
Formel: C₅H₁₂N₄O₃·H₂SO₄
MDL Number: MFCD00012618
CAS-nummer: 2219-31-0
EINECS: 218-728-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0% (calcd.on anh.substance)
Specific rotation [a]20/D +14.0 to +22.0 deg (C=2, H2O)
Water max. 7.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR