4-Brom-2-(trifluormethyl)phenol ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Bromo-2-hydroxybenzotrifluoride

B4493-5G B4493-25G
TCIAB4493-5GEA 321 DKK
TCIAB4493-5G TCIAB4493-25G
4-Brom-2-(trifluormethyl)phenol ≥97.0% (by GC)
4-Brom-2-(trifluormethyl)phenol
Formel: C₇H₄BrF₃O
MDL Number: MFCD05668981
CAS-nummer: 50824-04-9

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 83.0 to 87.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR