4,4'-Bis(5-hexyl-2-thienyl)-2,2'-bipyridyl ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4,4'-Bis(5-hexyl-2-thienyl)-2,2'-bipyridine, 4,4'-Bis(5-hexylthiophen-2-yl)-2,2'-bipyridine

B4420-200MG
TCIAB4420-200MGEA 764 DKK
TCIAB4420-200MG
4,4'-Bis(5-hexyl-2-thienyl)-2,2'-bipyridyl ≥98.0% (by HPLC, total nitrogen)
4,4'-Bis(5-hexyl-2-thienyl)-2,2'-bipyridyl
Formel: C₃₀H₃₆N₂S₂
MDL Number: MFCD22581259
CAS-nummer: 1047684-56-9

Bestil nu

Specifikationer Testresultater

Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR