(R)-1-Boc-3-Hydroxypiperidine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (R)-1-BOC-3-hydroxypiperidine, (R)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate

B4199-1G B4199-5G
TCIAB4199-1GEA 370 DKK
TCIAB4199-1G TCIAB4199-5G
(R)-1-Boc-3-Hydroxypiperidine ≥98.0% (by GC)
(R)-1-Boc-3-Hydroxypiperidine
Formel: C₁₀H₁₉NO₃
Molekylvægt: 201.27 g/mol
MDL Number: MFCD04115306
CAS-nummer: 143900-43-0

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C
Specific rotation [a]20/D -20.0 to -23.0 deg (C=5,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR